SP Bel-Art High Heat Minerit HD Desiccator Plates, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F42038-0230 F42038-0190 F42038-0140
80077-352CS 2913.15 CAD
80077-352 80077-356 80077-354
SP Bel-Art High Heat Minerit HD Desiccator Plates, Bel-Art Products, a part of SP
Desiccators Desiccators, Plates
These desiccator plates are able to withstand temperatures up to 150ºC (300ºF), have excellent resistance to thermal shock, are non-combustible, asbestos free, stable, and chemical-resistant.
  • Excellent thermal shock resistance
  • Chemical-resistant
  • Asbestos-free
  • Non-combustible
  • Space-saving design
Perforated with 6 mm (0.25") diameter holes on 25 mm (1") centers in a straight line pattern, they fit Scienceware® “Space Saver” desiccators, in addition to other plastic desiccators of similar size.