Synthware Three-Neck Round-Bottom Flasks with Bottom Outlet, Kemtech America
Supplier: Kemtech America
F424450 F42441L F42442L F42542L
60002-924EA 525.65 CAD
60002-924 60002-928 60002-934 60002-938
Synthware Three-Neck Round-Bottom Flasks with Bottom Outlet, Kemtech America
Flasks Round Bottom Flasks
Lower outlet is a 4mm straight bore PTFE stopcock.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Straight side necks
  • Bottom drain
  • Cap included
Side [ST] joints size: 24/40.