Synthware Round-Bottom Flasks with Threaded Inlet, Kemtech America
Supplier: Kemtech America
F141425 F141450 F149100 F144100 F141250 F149250 F144250 F144500
60002-372EA 92.49 CAD
60002-372 60002-374 60002-380 60002-382 60002-384 60002-386 60002-388 60002-390
Synthware Round-Bottom Flasks with Threaded Inlet, Kemtech America
Flasks Round Bottom Flasks
Features a threaded side port for insertion of thermometer or bleed tubes with an outside diameter between 5–7.5 mm.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Threaded side arm
  • Cap included
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.