HardyDisk™ Sulbactam-Durlobactam 10/10 μg (SUD20)
Supplier: HARDY DIAGNOSTICS CA
Z9461
Z9465
CA77497-270EA
61.11
CAD
CA77497-270
CA77497-272
HardyDisk™ Sulbactam-Durlobactam 10/10 μg (SUD20)
Microbiology Test Systems
Antibiotic Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.