Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
7120-0020 7120-0010
62662-886CS 702.7 CAD
62662-886 62662-875
Nalgene® Pans, High-Density Polyethylene, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans are constructed of high-density polyethylene for rigidity and durability
Use for soaking labware or as an ice bath. With handles for easy carrying. Tapered for nesting.