Primate Chairs, Plas-Labs™
Supplier: Plas-Labs
512-SPC/EXP 514-AG/EXP 515-SASR/EXP 517-CM/EXP 518-RPC/EXP 520-BS/EXP
CA89065-324EA 13120.19 CAD
CA89065-324 CA89065-326 CA89065-328 CA89065-330 CA89065-332 CA89065-334
Primate Chairs, Plas-Labs™
Animal Supplies Animal Cages
The rotating rhesus chair is designed for complete rotational studies of primates. Chairs feature horizontally and vertically adjustable restraint plates, food bars for self-feeding or dietary reward, anti-scuff casters with brakes and locks, heavy gauge optical quality acrylic walls, large hand knobs to prevent slipping of grip, and two seat styles for increased comfort.
  • Restraining portion rotates 360° on one set of uprights.
  • Ergonomic back plate supports
  • Tool-free locking and plastic-coated support floor
All surfaces susceptible to animal contactMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable