Instrument Trays, Stainless Steel, Polar ware
Supplier: Polar Ware
E1650 1202 E1654
62687-069EA 242.06 CAD
62687-069 62687-049 62687-070
Instrument Trays, Stainless Steel, Polar ware
Trays/Dishes/Pots Instrument Trays/Dishes
For sterilization of instruments, syringes, and more
Made of stainless steel for easy cleaning and quick sterilization. Stain-proof and long lasting — won’t chip, break, or corrode. May be ordered with or without solid flat covers. Same cover is used for trays 62687-069 and -070.