Synthware Distillation Receivers with Jacketed Uptake Tube Vacuum, Dean Stark, Kemtech America
Supplier: Kemtech America
D441405 D441410 D442410 D442420
60002-178EA 475.98 CAD
60002-178 60002-180 60002-186 60002-188
Synthware Distillation Receivers with Jacketed Uptake Tube Vacuum, Dean Stark, Kemtech America
Distillation Components Distillation Receivers
With jacketed uptake tube vacuum to improve distillation performance
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Graduated
  • Teflon® stopcock
  • Jacketed side arm
Jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. With 2mm PTFE stopcock.
Order Now
SPECIFICATIONS
Environmentally Preferable