SP Bel-Art Odo-Clave® Autoclave Deodorant Pads, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H13198-0003 H13199-0000 H13198-0001 H13198-0002 H13198-0000
39988-000PK 261.49 CAD
39988-000 11215-880 11215-983 11215-985 11215-987
SP Bel-Art Odo-Clave® Autoclave Deodorant Pads, Bel-Art Products, a part of SP
Deodorants and Air Fresheners
Heat-released deodorant pads neutralize autoclave odors with one of four pleasant scents.
  • Effectively neutralize autoclave odors
  • Choice of pleasant scents
  • Long lifespan, especially if stored properly
  • Easy to apply prior to autoclaving
Simply open foil pouch and place pad in bag prior to autoclaving
Also available in triple strength clove that releases three times the deodorant of the standard pads while costing less than two standard pads.
Pads have a three-year lifespan but will last longer if stored in a sealed container.
Order Now
SPECIFICATIONS
Environmentally Preferable