Cage Racks, Tecniplast®
Supplier: Tecniplast Usa
2D24B1011 2C30B1011 2E30B1021 2P15B1000
10716-312EA 13738.94 CAD
10716-312 95057-884 95057-890 10716-310 10716-315 77322-432
Cage Racks, Tecniplast®
Animal Supplies Animal Cages
Cage racks equipped with automatic watering equipment feature a Reverse-S manifold piping manufactured from corrosion-resistant stainless steel. Design permits excellent viewing of all cages (not included) and occupants, and improves hygiene, safety, and ergonomics.
  • Designed to allow positive air venting and complete water exchange in the manifold when flushing
  • Entire manifold assembly can be easily dismantled
  • Suspension-style racks minimize movement of individual cages when others are serviced or entire rack is moved, also eliminate bumping of adjacent filter covers
  • Constructed of stainless steel with nylon support rails
Shelf-style cage racks feature solid sheet stainless steel shelves for rigid, long-lasting performance.