VWR® Talon® Column Clamps
Supplier: VWR International
21572-104 21572-137 21572-160 21572-192
21572-104EA 172.33 CAD
21572-104 21572-137 21572-160 21572-192
VWR® Talon® Column Clamps
Clamps General Purpose Clamps
Sturdy, easily adjusted, multi-purpose clamps are ideal for holding large cylindrical glassware and similar objects.
  • Multi-purpose design
  • Sturdy
  • Easily adjusted
  • Corrosion-resistant stainless steel construction
A large, flat thumbscrew/worm-drive permits maximum tightening or removing in minimum time. Clamps are constructed of corrosion-resistant stainless steel. With 211 mm (85/16") extension arm.
Ordering information: An additional holder must be purchased in order to attach clamps to frames or other apparatus.