SP Bel-Art Universal Animal Restrainers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H46400-0003 H46400-0001 H46400-0002
47750-274EA 1007.71 CAD
47750-274 47750-270 47750-272
SP Bel-Art Universal Animal Restrainers, Bel-Art Products, a part of SP
Animal Supplies Animal Restraints
Clear acrylic plastic animal restrainers have removable head and tail gates.
  • Clear acrylic construction
  • Held together by chrome-plated brass thumb screws
  • Breathing holes to give air to animals
The head gate has breathing holes and fits into any of four positions for various sizes of rodents. The tail gate allows maximum closeness of occluding cuff to body. The inner raised floor may be removed for larger rodents. Knurled, chrome-plated brass thumb screws serve as base fasteners and legs.