VWR® Talon® Three-Prong Extension Clamps
Supplier: VWR
VWR® Talon®
915009 915007 915008 915850 915851 915006 915005 915004 915371 915852 915370
12621-238EA 187.39 CAD
12621-238 21570-200 97035-502 21570-126 12621-236 21570-302 21570-125 12621-240 21570-127 21570-404 97035-504
VWR® Talon® Three-Prong Extension Clamps
Clamps General Purpose Clamps
Clamps securely hold every type of glassware and apparatus.
  • Large grip adjustment range
  • Three-prong construction
  • Choose single or dual adjustment
  • Stainless steel or nickel-plated zinc
Long, seamless tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms that allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment. Stainless steel clamps offer exceptional chemical resistance, and are electropolished, made entirely of stainless steel, and autoclavable. Nickel-plated zinc offers a clamp with a high tensile strength at an economical price. Supplied with non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C.
Ordering information: An additional holder must be purchased in order to attach clamps to frames or other apparatus.