VWR® Talon® Two-Prong Extension Clamps
Supplier: VWR International
21570-007 21570-109 21570-115 21570-120 89202-612 89202-618
21570-007CS 2172.05 CAD
21570-007 21570-109 21570-115 21570-120 89202-612 89202-618
VWR® Talon® Two-Prong Extension Clamps
Clamps General Purpose Clamps
Extension arm attaches clamp head securely and offers easy positioning in the deepest fume hoods.
  • Large grip adjustment range
  • Two-prong construction
  • Single or dual adjustment
  • Stainless steel electropolished finish or nickel-plated zinc
Securely holds laboratory glassware and apparatus. Clamps are constructed with round extension arms that allow the clamps to be rotated 360°. Extension arms also allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment.
Ordering information: Supplied withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable