VWR® Talon® Two- and Three-Prong Swivel Clamps
Supplier: VWR International
21573-504 21573-606 21573-708 89202-622 89202-624
21573-504CS 2189.27 CAD
21573-504 21573-606 21573-708 89202-622 89202-624
VWR® Talon® Two- and Three-Prong Swivel Clamps
Clamps General Purpose Clamps
Swivel clamps hold apparatuses near the lab frame, and unlike extension clamps, the swivel clamps have an integral holder for attaching a lab frame or other apparatus.
  • electropolished stainless steel or nickel-plated zinc construction
  • For circular or irregular objects
  • Fully adjustable angle
  • Can be locked once in place
Built-in holder grips rods up to 19mm (3/4") in diameter and is adjustable for forward or reverse-facing adjustment screws. Shaft wing nut allows the holding angle of swivel clamp to adjust through 360° of rotation and can be locked in place once desired position is acheived. Stainless steel electropolished finish or nickel-plated zinc construction.
Ordering information: Supplied with non-slip vinyl sleeves, and for temperatures above 100°C (212°F), fiberglass covers.