Synthware Chromatography Columns with Beaded Top and PTFE Stopcock, Kemtech America
Supplier: Kemtech America
C361320 C361330 C361340 C361720 C361725 C361730 C362620 C362625 C362630 C362640 C363225 C364640 C366030 C366040 C366060 C368030
60001-638EA 161.05 CAD
60001-638 60001-640 60001-642 60001-644 60001-646 60001-648 60001-652 60001-654 60001-656 60001-658 60001-662 60001-674 60001-680 60001-682 60001-684 60001-690
Synthware Chromatography Columns with Beaded Top and PTFE Stopcock, Kemtech America
Chromatography Columns
Chromatography columns have a reinforced beaded top and PTFE stopcock.
  • Heavy wall Borosilicate construction
  • Beaded top
  • Teflon® stopcock
  • High capacity reservoir
Chromatography columns are used to separate out individual components of a liquid sample.