Synthware Chromatography Columns with Spherical Joint, Reservoir, and PTFE Stopcock
Supplier: Kemtech America
C381720 C381730 C381740 C382620 C382625 C382630 C382640 C383220 C383225 C383230 C383240 C384630 C384640
60001-538EA 223.27 CAD
60001-538 60001-540 60001-542 60001-544 60001-546 60001-548 60001-550 60001-552 60001-554 60001-556 60001-558 60001-564 60001-566
Synthware Chromatography Columns with Spherical Joint, Reservoir, and PTFE Stopcock
Chromatography Columns
Chromatography columns have a 35/20 spherical joint, reservoir, and PTFE stopcock.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Teflon® stopcock
  • High capacity reservoir
Chromatography columns are used to separate out individual components of a liquid sample.