Synthware Chromatography Columns with [ST] Joint, Reservoir, Fritted Disc, and PTFE Stopcock, Kemtech America
Supplier: Kemtech America
C391720C C391730C C391740C C392620C C392625C C392630C C392640C C393220C C393225C C393230C C393240C C394620C C394625C C394630C C394640C C396020C C396025C C396030C C396040C C396060C C398020C C398025C C398030C C398060C
60001-388EA 288.66 CAD
60001-388 60001-390 60001-392 60001-394 60001-396 60001-398 60001-400 60001-402 60001-404 60001-406 60001-408 60001-410 60001-412 60001-414 60001-416 60001-418 60001-420 60001-422 60001-424 60001-426 60001-428 60001-430 60001-432 60001-436
Synthware Chromatography Columns with [ST] Joint, Reservoir, Fritted Disc, and PTFE Stopcock, Kemtech America
Chromatography Columns
Chromatography columns have a [ST]24/40 outer joint at top, reservoir, coarse porosity sealed-in fritted disc to support column packing, and PTFE stopcock.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Teflon® stopcock
  • High capacity reservoir
  • Fritted disc
Sealed in fritted disc is not only perfect for filtration but also offers support for column packing.
Order Now
SPECIFICATIONS
Environmentally Preferable