Synthware Flash Chromatography Columns with [ST] Joint and PTFE Stopcock, Kemtech America
Supplier: Kemtech America
C184171 C184172 C184173 C184261 C184262 C184263 C184264 C184321 C184323 C184324 C184463 C184464 C184604
60001-238EA 240 CAD
60001-238 60001-240 60001-242 60001-244 60001-246 60001-248 60001-250 60001-252 60001-256 60001-258 60001-264 60001-266 60001-274
Synthware Flash Chromatography Columns with [ST] Joint and PTFE Stopcock, Kemtech America
Chromatography Columns
General purpose flash chromatography columns have a [ST] top outer joint and PTFE stopcock.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Teflon® stopcock
These columns are designed for flash chromatography.