Synthware Allihn Condensers with Removable Hose Connections, Kemtech America
Supplier: Kemtech America
C621120 C621200 C624300 C624400 C625300
60001-084EA 213.14 CAD
60001-084 60001-090 60001-092 60001-094 60001-102
Synthware Allihn Condensers with Removable Hose Connections, Kemtech America
Condensers
With joints at the top and bottom.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Allihn style condenser
Hose connection 8 mm (ext. Ø).