Gold Seal® Microscope Slides, Erie Scientific
Supplier: Erie Scientific
3032-002 3031-001
48312-172CS 2351.72 CAD
48312-172 48349-013 CA48312-068A CA48312-104 CA48312-160 CA48313-060
Gold Seal® Microscope Slides, Erie Scientific
Slides Microscope Slides
Gold Seal® slides are available in several popular styles
For additional sizes and thicknesses, contact your Avantor sales representative.
Double-frosted slides feature a frosting on both sides of one end. The rite-ON™ slides are frosted on one end with brilliant white epoxy ink. Slides that are frosted on one end or one side have a Gold Seal® logo printed key.