Synthware Coiled Condensers with Removable Hose Connections, Kemtech America
Supplier: Kemtech America
Synthware
C524200 C528300 C524400 C524300
60000-914EA 371.54 CAD
60000-914 60000-916 60000-912 60000-918
Synthware Coiled Condensers with Removable Hose Connections, Kemtech America
Condensers
With joints at top and bottom.
  • Nylon caps and hose connection included
  • Heavy wall Borosilicate construction
  • Ground glass joint
Hose connection O.D.: 8 mm.