Synthware Reflux Condensers with Removable Hose Connections, Kemtech America
Supplier: Kemtech America
C561100 C562280 C564125 C564175 C564225
60000-876EA 362.13 CAD
60000-876 60000-878 60000-882 60000-884 60000-886
Synthware Reflux Condensers with Removable Hose Connections, Kemtech America
Condensers
Coil-style reflux condensers with tightly wrapped coil for maximum cooling.
  • Nylon caps and hose connection included
  • Heavy wall borosilicate construction
  • Ground glass joint
Excellent for use with volatile solvents. Condensers feature a top outer [ST] joint and lower inner drip tip joint. Hose connections ext. Ø: 8 mm (at middle serration).