Airstrip Rubber Mats, Black, JCH International INC
Supplier: JCH International
2401 2402 2422G 2423G
56354-008CT 2803.36 CAD
56354-008 56355-000 56357-006 56358-009 56359-001
Airstrip Rubber Mats, Black, JCH International INC
Floor Mats Traction and Anti-Fatigue Floor Mats
Made of 1.3cm (1/2") thick black rubber with air pockets underneath to give buoyancy
Grease and chemical-resistant mats (NBR/SBR blend) also available.
Available as single sections (28x38") beveled on all sides to prevent tripping, or as interlocking sections (28x36") beveled on two sides. Two end sections interlock to make a 1.82m (6') length. Additional length is made up by center sections.