Stainless Steel Raised Wire Floors, Tecniplast®
Supplier: Tecniplast Usa
1290D150 2150E150 2154F150 1264C150
10714-105CS 2926.97 CAD
10714-105 77322-392 77322-358 77322-428 77322-378
Stainless Steel Raised Wire Floors, Tecniplast®
Animal Supplies Animal Cages
Stainless steel design with 30 mm elevation separates animals from bedding and waste.
Not required for standard operation but recommended for post-surgical animals or for those on restrictive diets. Help prevent foot pad swelling. Convenient handle design for easy insertion and removal. Autoclavable.