Synthware Bosshead, Kemtech America
Supplier: Kemtech America
W200002 W200002Z W200002B J111200
60005-036EA 42.54 CAD
60005-036 76264-412 76264-414 76264-416
Synthware Bosshead, Kemtech America
Clamps Bossheads
Chrome plated finish with plastic-headed screws.
  • Metal bosshead
  • Chrome plated and black metal available
  • Various tension screw designed available
Designed for use with rods ranging from 15 to 19 mm in diameter.