VWR® Disposable Aluminum Crinkle Dishes with Tabs
Supplier: VWR International
12577-081 16005-126 25433-008 25433-010 25433-052 25433-085
12577-081CS 366.59 CAD
12577-081 16005-126 25433-008 25433-010 25433-052 25433-085
VWR® Disposable Aluminum Crinkle Dishes with Tabs
Boats Weighing Boats
These dishes are ideal for general-purpose weighing, dispensing, and storing applications.
  • Dishes feature tabs for easy handling
  • Crimped sides for rigidity
  • Sturdy construction
Dishes also function as evaporating dishes, parts containers, or dust covers. Sturdy, with a flat bottom and slight taper for easy stacking.