VWR® Glass Joint Clamps, PTFE
Supplier: VWR International
89026-174 89026-180 89026-188
89026-174PK 162.23 CAD
89026-174 89026-180 89026-188
VWR® Glass Joint Clamps, PTFE
Clamps Standard Taper and Spherical Joint Clamps
PTFE joint clamps can withstand temperatures of up to 260°C (500°F).
  • Chemically resistant and inert
Feature PTFE-encapsulated steel springs that can be adjusted by hand.