Standard Taper Clamps, Wheaton
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
297746 297749 297752 297755 297757
21734-807CS 470.91 CAD
21734-807 21734-830 21734-862 21734-895 21734-920
Standard Taper Clamps, Wheaton
Clamps Standard Taper and Spherical Joint Clamps
Color-coded clamps hold conical ground joints secure under pressure.
Molded from acetal plastic, clamps are safe for use to 150°C (302°F). Unlike steel wire clamps, these will not scratch glass or bend out of shape. Available in the four most commonly used sizes; assorted pack 21734-920 includes six of each size/color.