Spilfyter® Waste Water Classifier, J.V. Mfg.
Supplier: FYTERTECH NONWOVENS LLC
580010 580050 581020 580001
15552-170CS 454.38 CAD
15552-170 15552-172 15552-174 56609-232 56609-256
Spilfyter® Waste Water Classifier, J.V. Mfg.
Environmental Test Kits
Distinguish risks in unknown wastewater batches and classify for proper treatment in a mater of seconds
Each wastewater classifier station (15552-174) includes 20 wastewater classifier strips, one wastewater classifier chart, two bobbins of polypropylene thread, and one weather-resistant plastic carry case. Each wastewater classifier kit (56609-232) includes six wastewater classifier strips, one wastewater classifier chart, and one bobbin of polypropylene thread. Additional strips and charts are available separately.
Strips simultaneously test for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks. Laminated wastewater classifier chart determines possible presence of wastewater chemical types according to color changes in strip.