Mouse Huts, Polycarbonate, Bio-Serv
Supplier: Bio-Serv
K3271 K3272 K3102
CA89067-846EA 32.03 CAD
CA89067-846 CA89067-848 CA89412-068
Mouse Huts, Polycarbonate, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Huts are sheltering devices for mice and other small rodents
Amber, red, or blue transparent polycarbonate allows easy visual checks, while the colored tint diffuses light, providing the mouse with a sense of security and protection. Cagewasher safe. Autoclavable.
Ideal for reducing anxiety, improving breeding, as well as a nesting place for dams and their litters. Low profile design allows it to fit into many different cage sizes without disturbing water sippers. Huts provide additional floor space since the animal can climb or rest on the flat, top surface. Open bottom prevents feces and urine accumulation.
Certifications: Certified (contaminant screened).