Dumbbell Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
K3227 K3224 K3223 K3270 K3227 K3224 K3223
CA89067-638EA 33.99 CAD
CA89067-638 CA89067-640 CA89067-642 CA89067-644 89067-638 89067-640 89067-642
Dumbbell Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Dumbbells provide animals with hours of gnawing, tossing, and other forms of enjoyment
Lightweight, easy-to-grip devices are made of long-lasting solid polypropylene. Large end knobs prevent slipping through cage bars. Certified (contaminant screened) and cage washer safe. Assorted bright colors. Autoclavable.
Ordering information: Recommended dumbbell sizes include medium, large, and X-large for dogs; small through X-large sizes for primates; medium and large sizes for rabbits; and small size for guinea pigs.