Max® and Max Lite® Earplugs, Honeywell Safety
Supplier: Honeywell Safety
Max® Max Lite®
LPF-30 MAX-1 MAX-30 MAX-1-D MAX-1-D LPF-30 MAX-30 LPF-1 MAX-1 LPF-1
CA55533-026CS 1021.75 CAD
55533-006 CA55533-026 CA55533-004 55533-026 55533-004 CA55533-002 55533-002 CA55533-008 55533-008 CA55533-006
Max® and Max Lite® Earplugs, Honeywell Safety
Earplugs Disposable Earplugs
Pre-shaped foam earplugs feature a smooth outer skin for maximum user comfort.
  • Disposable, non-irritating and non-allergenic
  • Available with or without connecting cord
  • Max Lite® low-pressure self-adjusting foam fits smaller ear canals
Certifications: Meet ANSI S3.19 standard. CE marked. CSA certified.
Ordering information: Earplugs 55533-026 are bulk-packed for use in the Howard Leight dispenser (see 89013-668 series).