Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Supplier: Thermo Fisher Scientific
700702
700402
700902
700001
700003
700003
34182-070CS
503.74
CAD
34182-070
34182-072
34182-074
34182-394
34182-587
34182-588
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Environmental Test Kits
Sample well water, rain water, distilled/deionized water, boiler feed water, process water, and more with the Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
- Increases measurement accuracy
- Reduces electrode drift
- Eliminates contamination of sample
- For best results, a high quality pH electrode is recommended
It offers fast, accurate pH results in high purity and low ionic strength waters.
Ordering information: Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing pH.