SP Wilmad-LabGlass Snyder Distilling Columns, SP Industries
Supplier: SP Industries
LG-5850-102 LG-5850-098
89003-962EA 409.81 CAD
89003-962 14205-014
SP Wilmad-LabGlass Snyder Distilling Columns, SP Industries
Distillation Components Distillation Columns
Columns include either two, three, or six sections designed for rapid solvent evaporation while retaining a high percentage of solute.
  • Borosilicate glass
They are used as a component in the Kuderna-Danish evaporative concentrator (LG-6867). Each section contains a floating ball valve which allows vapors to rise and condensate to flow downward.