KIMAX® Chromatography Columns with Reservoir, Straight Stopcock Assembly, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
17810-19300 17810-11300
89001-654CS 325.84 CAD
89001-654 21503-412
KIMAX® Chromatography Columns with Reservoir, Straight Stopcock Assembly, Kimble Chase
Chromatography Columns
These 300 mm columns feature a 250 ml reservoir and a reinforced top.
Lower end is tooled for extra strength and a secure, leakproof fit with the all-PTFE stopcock, ensuring that liquid contacts only glass and PTFE. An O-ring maintains tight seal over a 0 to 100 °C (32 to 212 °F) temperature range. Stopcock is easily removed for access to column packing.