KONTES® CHROMAFLEX™ Chromatography Columns, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420280-0232 420280-0213 420280-0222 420280-0242 420280-0252
KT420280-0232EA 487.26 CAD
KT420280-0232 KT420280-0213 KT420280-0222 KT420280-0242 KT420280-0252
KONTES® CHROMAFLEX™ Chromatography Columns, Kimble Chase
Chromatography Columns
General-purpose columns with an integral reservoir for analytical separations.
Constructed of borosilicate glass. Top is tooled for rubber stopper. A plug of glass wool can be used to support the column packing. Flow rate is controlled by a 2mm bore PTFE stopcock plug with an 11/25 plug size, a 1:5 taper, and a threaded pressure nut. Recommended for RCRA SW-846 methods.