BRAND PCR Plates, BrandTech®
Supplier: Brandtech
781400 781354 781357 781412 781416 781345 781350 781411 781415 781366 781368 781371 781373 781375 781377 781367 781369 781372 781374 781376 781378
80087-150PK 908.18 CAD
80087-150 89135-444 89135-446 89135-456 89135-458 80087-142 89135-438 89400-702 89400-712 89400-714 10141-426 10141-428 10141-430 10141-432 10141-434 10141-436 10141-438 10141-440 10141-442 10141-444 10141-446 10141-448
BRAND PCR Plates, BrandTech®
Microplates PCR Plates
Designed for applications requiring a high level of accuracy and reproducibility, these 24-well, 48-well, 96-well, and 384-well PCR plates are constructed from high-quality virgin plastics without the addition of lubricants or chemical additives that can inhibit reactions and contaminate samples
  • Ultra-thin walls for optimal heat transfer
  • Cleanroom quality
  • Models to fit popular thermal cyclers
Ultra-thin walled design facilitates constant, rapid, and precise heat transfer. Smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. Alphanumeric coding and cut corners are embossed and stamped in blue or black. Raised well rims protect against cross-contamination and provide a tight seal in combination with a matching sealing mat. White plastics for qPCR are uniformly colored with titanium dioxide (TiO2) to optimize reflection of the fluorescence signal. Autoclavable at 121°C, per DIN EN 285.
High quality sealing mats and films reduce evaporative losses during PCR process. Roller and cap tool enable easy sealing of films and caps with plates.
These PCR plates are suitable for use with most commercially available thermal cyclers, including low-profile instrumentation.
Certifications: DNase-, RNase-, and DNA-free.