BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Supplier: Brandtech
701346 701350 701352 701354 701355 701340 DW2200 DW2250
80087-064PK 1121.98 CAD
80087-064 80087-066 80087-068 80087-070 89093-076 76181-496 76183-224 76183-226
BRAND 96-Well and 384-Well Deep Well Plates, BrandTech
Microplates Deep Well Microplates
Deep well plates are excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, and sample storage
  • Suitable for storage, reagent reservoirs or use as mother plates
  • Work with most robotic/lquid handling systems
  • Polypropylene modes suitable for –80 °C (–112 °F) cryostorage
Polypropylene plates are manufactured from high-quality virgin polypropylene, are autoclavable to 121 °C (250 °F), and are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO. Plates are compatible with mostMore Product Information