KIMAX® Chromatographic Columns, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17800-22500 17800-11300 17800-11500 17800-19300 17800-19500 17800-22300
21503-156CS 361.38 CAD
21503-156 21503-010 21503-015 21503-065 21503-067 21503-123
KIMAX® Chromatographic Columns, Kimble Chase
Chromatography Columns
With removable stopcock of PTFE and replaceable glass tip.
Lower end of the column is tooled for extra strength and tight, leakproof fit. Glass tip is supplied.