Rodent Retreats Rat Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
K3246 K3245 K3170
CA89067-864EA 41.36 CAD
CA89067-864 CA89067-866 CA89412-098
Rodent Retreats Rat Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Polycarbonate devices offer rodents a place to rest comfortably on a solid surface, rather than on uncomfortable wire-bottom caging commonly used in many toxicology or other studies
Enhance animal well-being while minimizing stress and cage-associated foot lesions. Animals are easily observed through the transparent sides. Devices do not subtract from cage-space requirements and stack easily for cleaning. Cage washer-safe and autoclavable. May be used for aquatics. Certified (contaminant screened).
Order Now
SPECIFICATIONS
Environmentally Preferable