Synthware Distillation Heads, Jacketed Vigreux, Kemtech America
Supplier: Kemtech America
D211100 D211150 D214200 D214300
60002-144EA 704.66 CAD
60002-144 60002-146 60002-148 60002-150
Synthware Distillation Heads, Jacketed Vigreux, Kemtech America
Distillation Components Distillation Heads
Vacuum-jacketed Vigreux distillation heads feature a sealed-on condenser and are braced for strength
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Vigreux indentations
  • Designated 10/18 thermometer joint
Vacuum adapter has an inner joint and a sealed-in drip tip. Used with a 25mm immersion thermometer. Thermometer [ST] joint size: 10/18.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.