Reagent Bottles with Stoppers, Polypropylene, Narrow Mouth, BrandTech
Supplier: Brandtech
100494 100594 100394
83008-516PK 391.76 CAD
83008-516 83008-520 83008-512
Reagent Bottles with Stoppers, Polypropylene, Narrow Mouth, BrandTech
Bottles Plastic Bottles
Polypropylene reagent bottles with polypropylene stoppers provide excellent chemical resistance to many aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
  • Made of durable polypropylene
  • Tight fitting stoppers
  • Conical shoulders
Bottle 83008-532 has a handle. Bottles and stoppers are autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable