CiDehol® 70, Decon Labs
Supplier: Decon Labs
8401F 8432F 8416 8406 8405
CA11012-556CS 787 CAD
CA11012-556 CA11012-558 CA89125-206 CA89125-212 CA89125-214
CiDehol® 70, Decon Labs
Cleaning Agents Disinfectants
CiDehol® 70, 70% isopropyl alcohol (IPA) solution is made to USP specficiations and is filtered to 0.2µm to remove particulate contamination
It evaporates completely, with no rinsing required. Ideal for cleaning surfaces and hoods in microbiology labs or tissue culture labs. Effective in cleaning surfaces and process equipment in pharmaceutical, biotechnology, medical devices, cosmetic, and food production areas.