CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
8316F 8332F 8301F 8312
CA89125-194CS 743 CAD
CA89125-194 CA89125-196 CA89125-198 CA89176-254
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Cleaning Agents Disinfectants
Sterile, ready-to-use 70% isopropyl alcohol (IPA) solution is ideal for cleanroom use. Solution is filtered to 0.2 µm to remove particulate contamination. It evaporates completely, with no rinsing required.
Solution is filtered to 0.2 µm to remove particulate contamintion. It evaporates completely, with no rinsing required. Made according to USP specifications and packaged in a cleanroom. Each lot undergoes a bacterial endotoxin test to ensure that the product meets or exceeds Water-for-Injection quality in endotoxin limits, and each lot also undergoes a USP 14-day sterility test. Each case is shipped with lot specific documentation which details QC, irradiation, sterility, and bacterial endotoxin limits.
Ordering information: Each lot is double-bagged and gamma-irradiated to ensure sterility.