Isopropyl Alcohol Cleaner, Stoner
Supplier: Stoner
D005
89065-938EA 1038.39 CAD
89065-938
Isopropyl Alcohol Cleaner, Stoner
Cleaning Agents Detergents and Surface Cleansers
Pure isopropyl alcohol acts as a defluxer and general-purpose degreaser to remove dirt, oxides, oils, smoke film, and corrosion from metal and plastic surfaces.
  • IPA is 99.9% pure
  • Evaporates fast
  • Leaves no residue
Apply with squirt bottles, pump spray dispensers, swabs, wipes, or tank dipping techniques.