ESD Safe Spray Cleaner, R & R Lotion
Supplier: R & R Lotion
ICSC-GAL ICSC-16-ESD
56614-110CS 330.54 CAD
56614-110 56614-130
ESD Safe Spray Cleaner, R & R Lotion
Cleaning Agents Detergents and Surface Cleansers
ESD safe screen and workstation cleaner is designed for general-purpose use, and helps remove oils and dirt from surfaces without leaving streaks.
  • General-purpose, heavy-duty workstation cleaner
  • Biodegradeable, alcohol-free, and odorless
  • Can be used on terminals and flat panel displays
ESD Safe Spray Cleaner is suitable for use on terminals, keyboards, CRTs, and flat panel monitors
Work station cleaners can be used to help remove dust and dirt from computer terminals and other sensitive surfaces. Electrostatic discharge (ESD) is the sudden flow ofMore Product Information