Synthware Solvent Removal Vacuum Trap, Kemtech America
Supplier: Kemtech America
V120500 V121000
60004-714EA 313.89 CAD
60004-714 60004-716
Synthware Solvent Removal Vacuum Trap, Kemtech America
Vacuum Traps
Trap is designed for large volume solvent removal by vacuum.
  • 45/50 joint connects two components
  • Heavy wall borosilicate construction
With [ST] 40/35 joint. Center tube connection has an ext. Ø of 12 mm. The side tube connection has an ext. Ø of 10 mm.
Ordering information: Trap 60004-714 fits a 1900 ml Dewar flask. Trap 60004-716 fits a 4300 ml Dewar flask.