WOB-L® Dry Vacuum Pumps, Standard-Duty, Welch®
Supplier: Welch Vacuum
2561B-50 2534B-01 2546B-01 2511B-01 2511C-02 2522B-01
14230-628EA 5421.6 CAD
14230-628 14230-656 54970-544 54994-110 80077-612 80077-614 97000-076 97000-078 54994-106 80077-616
WOB-L® Dry Vacuum Pumps, Standard-Duty, Welch®
Pumps Vacuum Pumps
These oil-free pumps are ideal for basic lab operations.
  • Discharge air noise is muffled with a silencer
  • Not recommended for pumping acid, base, or organic vapors
Standard-duty pumps are protected from moisture and corrosion and use a WOB-L piston pumping mechanism. A liquid trap attached to each pump traps water if it is accidentally drawn into the line.
The 115 V, 60 Hz maintained on/off foot switch (14233-684, sold separately) allows hands-free operation of pumps. Made of durable black painted steel, the switch features a nonskid base and foot treadle pads to ensure secure footing during repeated operations. Comes with a 2.4 m (8') cord.
They are suited for applications requiring a rough vacuum of 648 to 752 mm Hg (251/2 to 2919/32" Hg) and free air displacement of up to 201 l/min. (7.1 cfm).
Ordering information: Chemical-duty models are also available, Contact your Avantor representative for more information.
Delivery information: Vacuum gauge included.