Rhino Series Refractometers, Percent Solids (Brix), Reichert
Supplier: Reichert Technologies
137531L0 137533L0
55680-347EA 787.71 CAD
55680-347 55680-369
Rhino Series Refractometers, Percent Solids (Brix), Reichert
Refractometers Handheld Refractometers
These automatic temperature-compensated hand refractometers are read directly and instantly without a water bath
Refractometers are constructed of durable, shock-resistant Xenoy® polycarbonate materials for demanding environments and applications. Water resistant. Dustproof.
No need to make separate temperature readings or refer to conversion tables. The Brix scale is based on percent by weight of sucrose. Useful in monitoring sugar level of soft drinks and juices and to check dissolved solids in ripening fruit crops.
Certifications: IP67 rated for entire unit, including eyepiece.
Order Now
SPECIFICATIONS
Environmentally Preferable
Accuracy ±1 scale division
Dimensions 23.5L x 3.8W x 3.8H cm (91/4 x 11/2 x 11/2")
Weight 162.4 g (5.7 oz.)